Learn More
Horst™ Rubber Insulating Mat
13.45€ - 18.95€
Specifications
Dimensions (L x W) | 2 Lm |
---|---|
Dimensions (L x W x H) | 2 Lm |
Product Type | Insulating Mat |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
10352082
|
Horst™
101706 |
25 mm |
18.95€
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
10168351
|
Horst™
101705 |
13 mm |
13.45€
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
Specifications
2 Lm | |
2 Lm | |
Insulating Mat |
Spot an opportunity for improvement?Share a Content Correction