Learn More
VACUUBRAND™ RZ 2.5 Rotary Vane Pump
Use for a wide range of laboratory and process applications that require low ultimate vacuum at medium gas flow rate Vacuubrand™ RZ 2.5 Rotary Vane Pump are high-performance rotary vane pumps with extra compact design and low weight.
Brand: VACUUBRAND™ 20698122
23501.94 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
High flow rates even at vacuum levels approaching ultimate vacuumHigh water vapor tolerance due to efficient gas ballast; very good ultimate vacuum even with gas ballast
Vacuum-tight at switch-off; external anti-suckback valve not needed large oil volume
Long intervals between oil changes ease of maintenance due to telescopic design
Specifications
1.5 x 10-3 torr (without Gas Ballast), 7.5 x 10-3 torr (with Gas Ballast) | |
UK Plug | |
CAN/CSA C22.2 61010-1-12, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, TUV, UL 61010A 61010-1:2012 | |
316 x 125 x 190 mm | |
43 dB(A) | |
104°F | |
12°C | |
50/60 Hz | |
314.96 mm | |
Rotary Vane Vacuum Pump | |
Chemical-Resistant | |
0.11 to 0.3 qt. | |
Oil-Sealed | |
16 psi |
0.18 hp | |
150 to 1800 rpm | |
230 V 50/60 Hz | |
RZ 2.5 | |
-10°C to +60°C | |
40°C | |
1.4/1.7 CFM | |
190.5 mm | |
124.46 mm | |
IP 40 | |
54°F | |
Laboratory | |
2 | |
230 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.