Learn More
SHOWA™ N-DEX™ 6005PF Disposable Nitrile Gloves
N-DEX™ 6005 series powder free gloves provide enhanced dexterity and performance across a range of tactical applications including food, electronics, janitorial, municipal services and utilities.
Especificaciones
Certificaciones/conformidad | 21 CFR 170-199 |
---|---|
Certificaciones/conformidad (Europa) | EN 455, Cat. I |
Aplicaciones | Industrial |
AQL | 1.5 |
Color | Azul |
Descripción
N-DEX™ 6005 series powder free gloves provide enhanced dexterity and performance across a range of tactical applications including food, electronics, janitorial, municipal services and utilities.
- Easy to put on and remove
- Can easily be one-hand used
- Ambidextrous
- Rolled cuff
- Smooth grip
- Made in the U.S.A.
- Specific applications include food handling, agriculture, washing and cleaning, public utility and coroner/morgue
Especificaciones
21 CFR 170-199 | |
Industrial | |
Azul | |
Sin textura | |
Nitrilo | |
4 mil | |
9,5 in |
EN 455, Cat. I | |
1.5 | |
Single-use | |
Sí | |
sin polvo | |
0.10 mm | |
24 cm |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Título del producto
SHOWA™ N-DEX™ 6005PF Disposable Nitrile Gloves