Learn More
Witeg™ Suction Bulb for Pipettes
A vacuum source for filling reagents and solutions.
Brand: Witeg™ 3968001
This item is not returnable.
View return policy
Description
- Green in color
- 1 mL capacity
- Provided 10 per pack
Specifications
Green | |
1 mL | |
Filling/transfer | |
Pipettes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction