Learn More
Sonoco™ ThermoSafe Aluminum Dollies for Storage /Transport Chests
Provide easy transport and handling for storage chests
Brand: Sonoco™ 376
This item is not returnable.
View return policy
Description
Provide easy transport and handling for storage chests
- Heavy-duty aluminum construction
- Four sturdy 4 in. (10cm) swivel casters with brakes
Specifications
Dolly | |
19 x 28.12 in. (48 x 71 cm) | |
Four sturdy swivel casters with brakes | |
48 cm | |
71 cm |
Aluminum | |
To provide easy transport and handling for storage chests | |
19 in. | |
28.12 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction