Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
503.40€ - 695.00€
Especificaciones
Tipo de poro | Mesh Pore Insert |
---|---|
Tipo de producto | Casete HistoScreen™ |
Código de producto | Marca | Color | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Color | Precio | Cantidad y disponibilidad | |||||
15363588
|
Epredia™
C-1000-AQ |
Aqua | N/A | ||||||
12658346
|
Epredia™
C-1000-BL |
Azul | N/A | ||||||
12668346
|
Epredia™
C-1000-GR |
Verde | N/A | ||||||
12678346
|
Epredia™
C-1000-GY |
Gris | N/A | ||||||
12688346
|
Epredia™
C-1000-LI |
Lila | N/A | ||||||
12698346
|
Epredia™
C-1000-OR |
Naranja | N/A | ||||||
12608356
|
Epredia™
C-1000-PK |
Rosa | N/A | ||||||
12618356
|
Epredia™
C-1000-TN |
Tan | N/A | ||||||
12628356
|
Epredia™
C-1000-WH |
Blanco | N/A | ||||||
12638356
|
Epredia™
C-1000-YL |
Yellow | N/A | ||||||
Descripción
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures tissues are safely processed as cassettes will not pop open during processing
Especificaciones
Mesh Pore Insert | |
Casete HistoScreen™ |