Learn More
TPPII Mouse anti-Human, CoraLite 594, Clone: 5H6F11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL594-66017-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The TPPII antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human TPPII. This antibody recognizes human antigen. The TPPII antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Especificaciones
TPPII | |
Monoclonal | |
CoraLite 594 | |
100 μL | |
7174 | |
IgG2b |
Immunofluorescence | |
5H6F11 | |
Mouse | |
Primary | |
Human |