Learn More
Tripropargilamina, 97 %, Thermo Scientific Chemicals
Código de producto. 15430298
Click to view available options
Quantità:
1 g
5 g
25 g
Tamaño de la unidad:
1 g
25 g
5 g
CAS: 6921-29-5 | C9H9N | 131.178 g/mol
Tripropargylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La tripropargilamina se utiliza como intermedio farmacéutico.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con los agentes oxidantes.
TRUSTED_SUSTAINABILITY
CAS 6921-29-5
Formula molecolare C9H9N
Molecular Weight (g/mol) 131.178
Numero MDL MFCD00008577
InChI Key ZHOBJWVNWMQMLF-UHFFFAOYSA-N
Sinonimo tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb
PubChem CID 23351
IUPAC Name N,N-bis(prop-2-inil)prop-2-in-1-amina
SMILES C#CCN(CC#C)CC#C
Specifications
Materiale o nome chimico Tripropargylamine
Densità 0.927
Punti di ebollizione 79°C to 85°C (11 mmHg)
Punto d'infiammabilità 57°C (135°F)
Formula lineare (HC≡CCH2)3N
Indice di rifrazione 1.484
Quantità 5 g
Numero UN UN1993
Peso formulazione 131.18
Percent Purity 97%
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.