Learn More
B Braun™ Sterican™ Single-use Needles
Manufactured from stainless chromium nickel-steel with transparent Luer Lock hub made of polypropylene. B Braun™ Sterican™ Single-use Needles have an extremely smooth surface with light silicone coating to provide minimal pain upon puncture.
Brand:  B Braun™ 9180109
Product Code. 15173683
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
View alternative products »
This item is not returnable. View return policy
Description
Description
  • „„Needle made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating „„for minimal pain upon puncture.
  • Transparent Luer Lock plastic hub made of polypropylene
  • „„Colour-coded hub identifies the outer diameter of the needle, conforms to ISO 6009
  • „„Latex-free/PVC-free
Specifications
Specifications
Sterile
EN ISO 7864 and DIN 13097
Green
Withdrawal and flushing
Polypropylene
22 mm
Stainless Chromium Nickel Steel
Luer Lock Hub
0.80 mm
21 G
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.