Learn More
Camlab™ Etiquetas de vinilo
Marca:  Camlab™ RTP095
Encompass_Preferred
Código de producto. 11712108
  • 36.44€ / paquete de 1000
Fecha estimada de envoi: 03-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Self adhesive, smear resistant with a high degree of chemical resistance
  • Available as Hi-Low strips and Hi-Low labels
  • Both types suitable for use between -85°C to 100°C
  • Designed for use with thermal transfer and laser printers
  • Hand feeding recommended
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.