Learn More
Thermo Scientific™ WebSeal™ Sealing Tapes
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes.
158.87 € - 929.63 €
Specifications
No. per Pack | 100 |
---|---|
Product Type | Non-sterile Sealing Tape |
Product Line | Webseal |
Adhesive | Pressure Sensitive |
Product Code | Brand | Material | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Color | Price | Quantity & Availability | ||||
15122719
|
Thermo Scientific™
60180-M142 |
Polyethylene, Silicone | Clear |
414.06 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15132719
|
Thermo Scientific™
60180-M143 |
PES, Silicone | Clear |
217.40 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15142719
|
Thermo Scientific™
60180-M144 |
Aluminum, Silicone | Silver |
158.87 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15152719
|
Thermo Scientific™
60180-M145 |
Polyolefin, Rubber | Clear |
294.47 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15162719
|
Thermo Scientific™
60180-M146 |
PET, Silicone | Clear |
929.63 €
Pack of 100 |
Estimated Shipment: 23-06-2025
Log in to see stock. |
||||
Description
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.
Easy, Consistent Sealing
- Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Tapes seal onto the plate with a convenient, handheld applicator (available separately)
Specifications
100 | |
Non-sterile Sealing Tape | |
Webseal | |
Pressure Sensitive |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.