Learn More
Euro Protection™ White SPP Polypropylene Lab Coats Without Pockets
112.00€
Specifications
Product Type | General Purpose Lab Coat |
---|---|
Gender | Unisex |
Disposable | Reusable |
Sterility | Non-sterile |
For Use With (Application) | Food, Industrial, Agriculture, Cleaning, Visitors |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12073206
|
Euro Protection™
41103 |
Large |
112.00€
Pack of 50 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
12083206
|
Euro Protection™
41104 |
X-Large |
112.00€
Pack of 50 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
12093206
|
Euro Protection™
41105 |
2X-Large |
112.00€
Pack of 50 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
Specifications
General Purpose Lab Coat | |
Reusable | |
Food, Industrial, Agriculture, Cleaning, Visitors |
Unisex | |
Non-sterile | |
40 g/m2 |
Spot an opportunity for improvement?Share a Content Correction