Learn More
WISP1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18166-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The WISP1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human WISP1. This antibody recognizes human, mouse antigen. The WISP1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
WISP1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
8840 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
433 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |