Learn More
Ansell AlphaTec™ 4000 Ultrasonically Welded and Taped Suit
AlphaTec™ 4000 Ultrasonically Welded and Taped Suit with innovative multi-layer chemical barrier technology and Type 3/4/5 protection.
Brand: Ansell OR40T-00130-09
661.38 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
- Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
- Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
- Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
- Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
- Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
- CE Category III Type 3/4/5 protection
Specifications
Orange | |
Multi-layer Barrier Laminate Fabric | |
Elasticated | |
3-piece hood | |
Ultrasonicaly welded and taped |
Attached | |
5X-Large | |
Single zip closure | |
Elasticated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction