Learn More
CPLX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16949-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Positively regulates a late step in synaptic vesicle exocytosis.Especificaciones
CPLX3 | |
Polyclonal | |
Unconjugated | |
CPLX3 | |
complexin 3, Complexin III, CPLX3, CPX III, CPXIII, Nbla11589 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235415, 501005, 594855 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R1B5, Q8WVH0 | |
CPLX3 | |
CPLX3 Fusion Protein Ag10571 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |