Learn More
Methyl 6-bromopyridine-2-carboxylate, 97%, Thermo Scientific Chemicals
397.00 NOK - 1267.67 NOK
Chemical Identifiers
CAS | 26218-75-7 |
---|---|
Molecular Formula | C7H6BrNO2 |
Molecular Weight (g/mol) | 216.03 |
MDL Number | MFCD06203934 |
InChI Key | SGNCOKUHMXLGAH-UHFFFAOYSA-N |
Synonym | methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate |
PubChem CID | 16217886 |
IUPAC Name | methyl 6-bromopyridine-2-carboxylate |
SMILES | COC(=O)C1=NC(Br)=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26218-75-7 | |
216.03 | |
SGNCOKUHMXLGAH-UHFFFAOYSA-N | |
16217886 | |
COC(=O)C1=NC(Br)=CC=C1 |
C7H6BrNO2 | |
MFCD06203934 | |
methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate | |
methyl 6-bromopyridine-2-carboxylate |
Specifications
26218-75-7 | |
White to Yellow | |
96% min. (HPLC) | |
C7H6BrNO2 | |
methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate | |
COC(=O)C1=NC(Br)=CC=C1 | |
216.03 | |
216.03 | |
Crystalline Powder or Needles |
94.6°C to 95°C | |
Authentic | |
Glass Bottle | |
MFCD06203934 | |
SGNCOKUHMXLGAH-UHFFFAOYSA-N | |
methyl 6-bromopyridine-2-carboxylate | |
16217886 | |
97% | |
Methyl 6-bromopyridine-2-carboxylate |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
- Methyl 6-bromopyridine-2-carboxylate, 97%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.