Learn More
BUCHI Rotavapor R-300 Vertical Condenser Glass Assembly
Vertical Condenser Glass Assembly for Rotavapor R-300, 1500 cm2. Assembly features a large top inlet for easy cleaning, connection for vapor temperature sensor, and inner glass tubing/plate. Includes a 1 liter receiving flask, tubing, and a ball joint clamp.
Marca: BUCHI 11062432
1423.33 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
- Includes receiving flask (1 L), tubing, and a ball joint clamp
- Does NOT include evaporating flask, vacuum seal, vapor duct, and condenser holder
- For use with Rotavapor R-300, R-2XX
Especificaciones
Borosilicate Glass (3.3) | |
Rotavap™ R-300, R-210, and R-215 | |
No Safety Coating |
Vertical Condensor | |
Receiving Flask, Tubing, and a Ball Joint Clamp | |
1 L |