Learn More
Berkshire Super Polx™ SWSE
Super Polx™ SWSE is a sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments.
Marca: Berkshire SPSWSE121210
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- 100% continuous filament double knitted polyester
- Ultrasonically sealed edge
- Laundered and packaged in an ISO Class 4 (Class 10) cleanroom
- Lot tracing on each pack
- Economical
- Strong, tear and abrasion resistant
- Very good absorbency
- Low particles and fibers
- Soft hand and edge for sensitive surfaces
Especificaciones
Surface Wiper | |
30 cm | |
30 cm | |
No esterilizable en autoclave | |
1-ply |
11.8 in. | |
12 in | |
Poliéster | |
Un solo uso | |
No estéril |