Learn More
Gilson™ Digital Dry Bath Block
Accessories for the Digital Dry Bath series.
184.00 €
Specifications
For Use With (Equipment) | Gilson Digital Dry Bath Series |
---|---|
Type | Block for Digital Dry Bath |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
16388621
|
Gilson™
36117200 |
5 x 50 mL centrifuge tubes |
184.00 €
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
|||||
16348621
|
Gilson™
36117160 |
12 x 5.0 mL centrifuge tubes (17 mm) |
184.00 €
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
16358621
|
Gilson™
36117170 |
12 x 15 mL centrifuge tubes |
184.00 €
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
16398621
|
Gilson™
36117210 |
12 x 15 mm or 16 mm test tubes |
184.00 €
Each |
Estimated Shipment: 10-10-2025
Log in to see stock. |
|||||
16368621
|
Gilson™
36117180 |
20 x 10 mm test tubes or 20 x 2 mL centrifuge tubes |
184.00 €
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
16338621
|
Gilson™
36117150 |
24 x 0.5 mL centrifuge tubes |
184.00 €
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
16318621
|
Gilson™
36117130 |
24 x 1.5 mL centrifuge tubes, conical |
184.00 €
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
|||||
16328621
|
Gilson™
36117140 |
24 x 1.5 mL or 2.0 mL centrifuge tubes |
184.00 €
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
16308621
|
Gilson™
36117120 |
48 x 0.2 mL test tubes or 6 PCR strips, 8 tubes each |
184.00 €
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
Description
Accessory for Digital Dry Bath SeriesSpecifications
Gilson Digital Dry Bath Series | |
Block for Digital Dry Bath |
Spot an opportunity for improvement?Share a Content Correction